Assay Details
| Gene Symbol: FAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005245.3 | NP_005236.2 | 13 - 14 | 9652 | 73 | |
| XM_005262834.2 | XP_005262891.1 | 13 - 14 | 10044 | 73 | ||
| XM_005262835.2 | XP_005262892.1 | 13 - 14 | 9669 | 73 | ||
| XM_006714139.2 | XP_006714202.1 | 13 - 14 | 10044 | 73 | ||
| GenBank mRNA | CN289962.1 | - | 3 - 4 | 440 | 73 | |
| X87241.1 | - | 13 - 14 | 9656 | 73 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001824.hg.1
Set Membership: |
|

FAT1
Genomic Map