Assay Details
| Gene Symbol: FAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005245.3 | NP_005236.2 | 15 - 16 | 10252 | 82 | |
| XM_005262834.2 | XP_005262891.1 | 15 - 16 | 10644 | 82 | ||
| XM_005262835.2 | XP_005262892.1 | 15 - 16 | 10269 | 82 | ||
| XM_006714139.2 | XP_006714202.1 | 15 - 16 | 10644 | 82 | ||
| GenBank mRNA | X87241.1 | - | 15 - 16 | 10256 | 82 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001824.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
actin filament organization
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
establishment or maintenance of cell polarity
cell-cell signaling
anatomical structure morphogenesis
single organismal cell-cell adhesion
cell migration
calcium ion binding
protein binding
actin filament organization
cell adhesion
homophilic cell adhesion via plasma membrane adhesion molecules
establishment or maintenance of cell polarity
cell-cell signaling
anatomical structure morphogenesis
single organismal cell-cell adhesion
cell migration
calcium ion binding
protein binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000441802.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000500085.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000503253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000503253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000503253.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507105.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000507662.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000508035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000508035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000508035.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509537.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509647.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000509927.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512347.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000512772.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | ENST00000614102.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012670.hg.1 | NM_005245.3 | Refseq |

FAT1
Genomic Map