Assay Details
| Gene Symbol: FAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005245.3 | NP_005236.2 | 18 - 19 | 10735 | 78 | |
| XM_005262834.2 | XP_005262891.1 | 18 - 19 | 11127 | 78 | ||
| XM_005262835.2 | XP_005262892.1 | 18 - 19 | 10752 | 78 | ||
| XM_006714139.2 | XP_006714202.1 | 18 - 19 | 11127 | 78 | ||
| GenBank mRNA | X87241.1 | - | 18 - 19 | 10739 | 78 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001824.hg.1
Set Membership: |
|

FAT1
Genomic Map