Assay Details
| Gene Symbol: FAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_005245.3 | NP_005236.2 | 4 - 5 | 3832 | 88 | |
| XM_005262834.2 | XP_005262891.1 | 4 - 5 | 4224 | 88 | ||
| XM_005262835.2 | XP_005262892.1 | 4 - 5 | 3849 | 88 | ||
| XM_006714139.2 | XP_006714202.1 | 4 - 5 | 4224 | 88 | ||
| GenBank mRNA | X87241.1 | - | 4 - 5 | 3830 | 88 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001824.hg.1
Set Membership: |
|

FAT1
Genomic Map