Assay Details
| Gene Symbol: STXBP5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001127715.2 | NP_001121187.1 | 6 - 7 | 807 | 148 | |
| NM_139244.4 | NP_640337.3 | 6 - 7 | 807 | 148 | ||
| XM_006715339.2 | XP_006715402.1 | 6 - 7 | 981 | 148 | ||
| XM_017010268.1 | XP_016865757.1 | 6 - 7 | 844 | 148 | ||
| GenBank mRNA | AL834152.1 | - | 6 - 7 | 793 | 148 | |
| BC107737.1 | - | 6 - 7 | 744 | 148 | ||
| BC113382.1 | - | 6 - 7 | 680 | 8322434 | 148 | |
| BC113408.1 | - | 6 - 7 | 680 | 8322460 | 148 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC06001071.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
exocytosis
regulation of gene expression
protein transport
regulation of exocytosis
regulation of blood coagulation
positive regulation of GTPase activity
positive regulation of exocytosis
regulation of protein secretion
GTPase activator activity
syntaxin-1 binding
Rab GTPase binding
syntaxin binding
exocytosis
regulation of gene expression
protein transport
regulation of exocytosis
regulation of blood coagulation
positive regulation of GTPase activity
positive regulation of exocytosis
regulation of protein secretion
GTPase activator activity
syntaxin-1 binding
Rab GTPase binding
syntaxin binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000321680.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367475.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367480.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000367481.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000392291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000392291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000392291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000392291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000392291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000392291.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000443556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000443556.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | ENST00000546097.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_001127715.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0600009753.hg.1 | NM_139244.4 | Refseq |

STXBP5
Genomic Map