| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037809.hg.1 |
TC01002456.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037809.hg.1 |
ENST00000271069.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037809.hg.1 |
ENST00000373672.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR01037809.hg.1 |
NM_001856.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000373672.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000468459.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000479100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000488128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
NM_001856.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000373672.7 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000468459.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000479100.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
ENST00000488128.6 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
NM_001856.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0100200773.hg.1 |
TC0100013576.hg.1 |
Clariom D TC |