Assay Details
| Gene Symbol: LOC100288798 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_125381.1 | 1 - 2 | 64 | |||
| GenBank mRNA | AI094227.1 | - | 2 - 3 | 416 | 64 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000551503.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | ENST00000607353.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125377.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125378.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125379.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125380.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200012618.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | ENST00000551021.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | NR_125381.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200199715.hg.1 | TC1200012618.hg.1 | Clariom D TC |

LOC100288798
Genomic Map