Assay Details
| Gene Symbol: LSAMP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001318915.1 | NP_001305844.1 | 8 - 9 | 1502 | 67 | |
| XM_017006384.1 | XP_016861873.1 | 7 - 8 | 724 | 67 | ||
| GenBank mRNA | AK299851.1 | - | 1317 | 67 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000333617.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000473171.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000474851.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000475403.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000490035.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000498645.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | ENST00000539563.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0300012073.hg.1 | NM_002338.4 | Refseq |

LSAMP
Genomic Map