| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600007655.hg.1 |
ENST00000570988.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010126.hg.1 |
ENST00000567803.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
TC1600010126.hg.1 |
ENST00000567803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010126.hg.1 |
ENST00000567803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600010126.hg.1 |
ENST00000567803.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007655.hg.1 |
ENST00000319817.3 |
Ensembl |
| Affymetrix Human Clariom D Assay |
TC1600007655.hg.1 |
ENST00000570988.1 |
Ensembl |