Assay Details
| Gene Symbol: ULK4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001322500.1 | NP_001309429.1 | 26 - 27 | 3156 | 136 | |
| NM_001322501.1 | NP_001309430.1 | 25 - 26 | 3080 | 136 | ||
| NM_017886.3 | NP_060356.2 | 26 - 27 | 3156 | 136 | ||
| XM_011533874.2 | XP_011532176.1 | 26 - 27 | 3734 | 136 | ||
| XM_011533880.2 | XP_011532182.1 | 10 - 11 | 1751 | 136 | ||
| XM_017006711.1 | XP_016862200.1 | 25 - 26 | 3650 | 136 | ||
| XM_017006712.1 | XP_016862201.1 | 26 - 27 | 3731 | 136 | ||
| GenBank mRNA | AK093396.1 | - | 5 - 6 | 408 | 136 | |
| AK304341.1 | - | 26 - 27 | 2787 | 136 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC03001312.hg.1
Set Membership: |
|

ULK4
Genomic Map