Assay Details
| Gene Symbol: LINC01109 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| GenBank mRNA | AK124684.1 | - | 1 | 657 | 138 | |
Adjacent Gene Details
| Gene name | Gene Symbol | Alias | Distance | Relative Direction | Location Chromsome | |
|---|---|---|---|---|---|---|
| Upstream | hepatocyte nuclear factor 4 gamma | HNF4G |
NR2A2, NR2A3 |
-837.5kb | Sense | Chr. 8 - 75407567 - 75566834 on NCBI Build GRCh38 |
| Downstream | long intergenic non-protein coding RNA 1111 | LINC01111 | 1.8kb | Sense | Chr. 8 - 76406654 - 76524356 on NCBI Build GRCh38 |
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect genomic DNA. Please contact Tech Support if you need more information.
Set Membership: |
|

LINC01109
Genomic Map