Assay Details
Gene Symbol: E2F3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005248866.4 | XP_005248923.1 | 1 - 2 | 947 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_005248866.4 transcripts.
Set Membership: |
![]() ![]() ![]() |
Panther Classification:
Molecular Function -



Gene Ontology Categories:
