Assay Details
Gene Symbol: AGO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001317122.1 | NP_001304051.1 | 10 - 11 | 1491 | 66 | |
NM_001317123.1 | NP_001304052.1 | 10 - 11 | 1335 | 66 | ||
NM_012199.4 | NP_036331.1 | 10 - 11 | 1491 | 66 | ||
XM_017001017.1 | XP_016856506.1 | 10 - 11 | 1452 | 66 | ||
XM_017001019.1 | XP_016856508.1 | 10 - 11 | 1549 | 66 | ||
GenBank mRNA | AF093097.1 | - | 10 - 11 | 1477 | 66 | |
AK001622.1 | - | 10 - 11 | 1335 | 66 | ||
AK314148.1 | - | 10 - 11 | 1452 | 66 | ||
BC063275.1 | - | 10 - 11 | 1471 | 30344513 | 66 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01000463.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
nuclear-transcribed mRNA catabolic process
transcription, DNA-templated
Wnt signaling pathway, calcium modulating pathway
RNA secondary structure unwinding
miRNA metabolic process
gene silencing by RNA
pre-miRNA processing
posttranscriptional gene silencing by RNA
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
mRNA cleavage involved in gene silencing by miRNA
miRNA loading onto RISC involved in gene silencing by miRNA
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol-mediated signaling
mRNA cleavage involved in gene silencing by siRNA
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core binding
core promoter binding
RNA binding
double-stranded RNA binding
single-stranded RNA binding
endoribonuclease activity
protein binding
miRNA binding
poly(A) RNA binding
nuclear-transcribed mRNA catabolic process
transcription, DNA-templated
Wnt signaling pathway, calcium modulating pathway
RNA secondary structure unwinding
miRNA metabolic process
gene silencing by RNA
pre-miRNA processing
posttranscriptional gene silencing by RNA
production of miRNAs involved in gene silencing by miRNA
miRNA mediated inhibition of translation
mRNA cleavage involved in gene silencing by miRNA
miRNA loading onto RISC involved in gene silencing by miRNA
positive regulation of transcription from RNA polymerase II promoter
phosphatidylinositol-mediated signaling
mRNA cleavage involved in gene silencing by siRNA
RNA polymerase II core promoter proximal region sequence-specific DNA binding
RNA polymerase II core binding
core promoter binding
RNA binding
double-stranded RNA binding
single-stranded RNA binding
endoribonuclease activity
protein binding
miRNA binding
poly(A) RNA binding
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | ENST00000624357.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01007141.hg.1 | TC01000463.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01007141.hg.1 | ENST00000373204.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007141.hg.1 | ENST00000373206.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01007141.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | ENST00000624357.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | TC0100007786.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | ENST00000373204.4 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | ENST00000373206.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | NM_012199.4 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100154817.hg.1 | TC0100007786.hg.1 | Clariom D TC |
