Assay Details
| Gene Symbol: PIFO | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181643.5 | NP_857594.2 | 2 - 3 | 255 | 99 | |
| XM_005270472.1 | XP_005270529.1 | 2 - 3 | 364 | 99 | ||
| GenBank mRNA | AK074433.1 | - | 2 - 3 | 190 | 99 | |
| BC029237.1 | - | 1 - 2 | 144 | 99 | ||
| BC050319.1 | - | 2 - 3 | 219 | 4837146 | 99 | |
| BC068603.1 | - | 2 - 3 | 179 | 99 | ||
| BC093891.1 | - | 2 - 3 | 242 | 7939736 | 99 | |
| BC101501.1 | - | 2 - 3 | 243 | 8069007 | 99 | |
| HM237139.1 | - | 2 - 3 | 242 | 99 | ||
| HM237140.1 | - | 2 - 3 | 168 | 99 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01000975.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR01015566.hg.1 | TC01000975.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015566.hg.1 | ENST00000369738.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015566.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR01015566.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0100167730.hg.1 | TC0100009408.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369737.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000369738.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000468395.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | ENST00000484512.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_001300831.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NM_181643.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0100009408.hg.1 | NR_125338.1 | Refseq |

PIFO
Genomic Map