Assay Details
| Gene Symbol: R3HDM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538031.1 | XP_011536333.1 | 2 - 3 | 269 | 80 | |
| XM_011538033.1 | XP_011536335.1 | 2 - 3 | 269 | 80 | ||
| XM_011538035.1 | XP_011536337.1 | 2 - 3 | 269 | 80 | ||
| XM_011538036.1 | XP_011536338.1 | 2 - 3 | 269 | 80 | ||
| XM_011538038.1 | XP_011536340.1 | 2 - 3 | 269 | 80 | ||
| XM_011538039.1 | XP_011536341.1 | 2 - 3 | 269 | 80 | ||
| XM_011538040.1 | XP_011536342.1 | 2 - 3 | 269 | 80 | ||
| XM_011538042.2 | XP_011536344.1 | 3 - 4 | 365 | 80 | ||
| XM_017018999.1 | XP_016874488.1 | 3 - 4 | 406 | 80 | ||
| XM_017019000.1 | XP_016874489.1 | 2 - 3 | 269 | 80 | ||
| XM_017019003.1 | XP_016874492.1 | 2 - 3 | 269 | 80 | ||
| XM_017019004.1 | XP_016874493.1 | 2 - 3 | 269 | 80 | ||
| XM_017019008.1 | XP_016874497.1 | 2 - 3 | 269 | 80 | ||
| XM_017019009.1 | XP_016874498.1 | 2 - 3 | 269 | 80 | ||
| XM_017019011.1 | XP_016874500.1 | 4 - 5 | 422 | 80 | ||
| XM_017019012.1 | XP_016874501.1 | 2 - 3 | 269 | 80 | ||
| XM_017019019.1 | XP_016874508.1 | 3 - 4 | 363 | 80 | ||
| XM_017019020.1 | XP_016874509.1 | 2 - 3 | 269 | 80 | ||
| XM_017019026.1 | XP_016874515.1 | 3 - 4 | 348 | 80 | ||
More Information
Set Membership: |
|

R3HDM2
Genomic Map