Assay Details
| Gene Symbol: ACO2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098.2 | NP_001089.1 | 4 - 5 | 546 | 74 | |
| XM_017028812.1 | XP_016884301.1 | 4 - 5 | 860 | 74 | ||
| GenBank mRNA | AK296260.1 | - | 5 - 6 | 511 | 74 | |
| AK297213.1 | - | 4 - 5 | 421 | 74 | ||
| AK302691.1 | - | 5 - 6 | 496 | 74 | ||
| AK314845.1 | - | 4 - 5 | 554 | 74 | ||
| BC014092.2 | - | 4 - 5 | 546 | 4328775 | 74 | |
| BC026196.2 | - | 4 - 5 | 549 | 5264179 | 74 | |
| CR456365.1 | - | 4 - 5 | 538 | 74 | ||
| CR536568.1 | - | 4 - 5 | 524 | 74 | ||
| GQ472196.1 | - | 4 - 5 | 549 | 74 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC22000337.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006502.hg.1 | ENST00000216254.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006502.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006502.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006502.hg.1 | ENST00000544234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006502.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006502.hg.1 | TC22000337.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006499.hg.1 | ENST00000216254.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006499.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006499.hg.1 | ENST00000544234.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006499.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR22006499.hg.1 | TC22000337.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | TC2200007477.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152849.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | TC2200007477.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000481310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | ENST00000481310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR2200152852.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000216254.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000396512.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000466237.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000471094.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000478010.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000481310.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | ENST00000482208.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC2200007477.hg.1 | NM_001098.2 | Refseq |

ACO2
Genomic Map