Assay Details
| Gene Symbol: EMP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_006720864.3 | XP_006720927.1 | 1 - 2 | 163 | 131 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_006720864.3 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
membrane raft assembly
T cell mediated cytotoxicity
positive regulation of cell-matrix adhesion
regulation of glomerular filtration
actin filament organization
cell adhesion
cell-matrix adhesion
embryo implantation
cell death
cell proliferation
positive regulation of cell proliferation
regulation of endothelial cell migration
cell migration
bleb assembly
activation of protein kinase activity
protein localization to cell surface
blood vessel endothelial cell migration
regulation of kinase activity
early endosome to late endosome transport
regulation of angiogenesis
actin-mediated cell contraction
caveola assembly
protein localization to plasma membrane
positive regulation of integrin-mediated signaling pathway
regulation of vasculogenesis
integrin binding
protein binding
kinase binding
protein kinase binding
membrane raft assembly
T cell mediated cytotoxicity
positive regulation of cell-matrix adhesion
regulation of glomerular filtration
actin filament organization
cell adhesion
cell-matrix adhesion
embryo implantation
cell death
cell proliferation
positive regulation of cell proliferation
regulation of endothelial cell migration
cell migration
bleb assembly
activation of protein kinase activity
protein localization to cell surface
blood vessel endothelial cell migration
regulation of kinase activity
early endosome to late endosome transport
regulation of angiogenesis
actin-mediated cell contraction
caveola assembly
protein localization to plasma membrane
positive regulation of integrin-mediated signaling pathway
regulation of vasculogenesis
integrin binding
protein binding
kinase binding
protein kinase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200012027.hg.1 | ENST00000443818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200012027.hg.1 | ENST00000443818.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000342147.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000359543.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | ENST00000536829.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1600009362.hg.1 | NM_001424.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200012027.hg.1 | ENST00000443818.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200012027.hg.1 | ENST00000443818.1 | Ensembl |

EMP2
Genomic Map