Assay Details
| Gene Symbol: ACADL | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001608.3 | NP_001599.1 | 1 - 2 | 304 | 101 | |
| GenBank mRNA | AK296161.1 | - | 1 - 2 | 261 | 101 | |
| AK313498.1 | - | 1 - 2 | 258 | 101 | ||
| BC039063.1 | - | 1 - 2 | 249 | 5189329 | 101 | |
| BC064549.1 | - | 1 - 2 | 236 | 5745188 | 101 | |
| DC350098.1 | - | 1 - 2 | 304 | 101 | ||
| M74096.1 | - | 1 - 2 | 81 | 101 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02002731.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
temperature homeostasis
fatty acid beta-oxidation
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
carnitine catabolic process
long-chain fatty acid catabolic process
cellular lipid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
protein homotetramerization
lipid homeostasis
oxidation-reduction process
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
long-chain-acyl-CoA dehydrogenase activity
electron carrier activity
palmitoyl-CoA oxidase activity
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor
temperature homeostasis
fatty acid beta-oxidation
carnitine metabolic process, CoA-linked
fatty acid beta-oxidation using acyl-CoA dehydrogenase
carnitine catabolic process
long-chain fatty acid catabolic process
cellular lipid catabolic process
negative regulation of fatty acid biosynthetic process
negative regulation of fatty acid oxidation
protein homotetramerization
lipid homeostasis
oxidation-reduction process
regulation of cholesterol metabolic process
fatty-acyl-CoA binding
acyl-CoA dehydrogenase activity
long-chain-acyl-CoA dehydrogenase activity
electron carrier activity
palmitoyl-CoA oxidase activity
flavin adenine dinucleotide binding
oxidoreductase activity, acting on the CH-CH group of donors, with a flavin as acceptor
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043858.hg.1 | TC02002731.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043858.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043858.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043856.hg.1 | TC02002731.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043856.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043856.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043861.hg.1 | TC02002731.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043861.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02043861.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214452.hg.1 | TC0200015603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214453.hg.1 | TC0200015603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214450.hg.1 | TC0200015603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200214456.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214456.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214456.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200214456.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200214456.hg.1 | TC0200015603.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000233710.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482502.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000482523.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | ENST00000498120.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200015603.hg.1 | NM_001608.3 | Refseq |

ACADL
Genomic Map