Assay Details
| Gene Symbol: L2HGDH | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024884.2 | NP_079160.1 | 6 - 7 | 814 | 151 | |
| XM_005268075.4 | XP_005268132.1 | 6 - 7 | 814 | 151 | ||
| XM_011537166.2 | XP_011535468.1 | 7 - 8 | 921 | 151 | ||
| XM_011537167.2 | XP_011535469.1 | 6 - 7 | 1199 | 151 | ||
| XM_011537168.2 | XP_011535470.1 | 4 - 5 | 433 | 151 | ||
| XM_017021655.1 | XP_016877144.1 | 6 - 7 | 945 | 151 | ||
| XM_017021656.1 | XP_016877145.1 | 7 - 8 | 899 | 151 | ||
| XM_017021657.1 | XP_016877146.1 | 7 - 8 | 826 | 151 | ||
| GenBank mRNA | AK022680.1 | - | 6 - 7 | 814 | 151 | |
| AK292627.1 | - | 6 - 7 | 814 | 151 | ||
| AY757363.1 | - | 6 - 7 | 749 | 151 | ||
| BC006117.1 | - | 6 - 7 | 767 | 3504996 | 151 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001117.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR14012597.hg.1 | ENST00000261699.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012597.hg.1 | ENST00000267436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012597.hg.1 | ENST00000421284.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012597.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012597.hg.1 | TC14001117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012596.hg.1 | ENST00000261699.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012596.hg.1 | ENST00000267436.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012596.hg.1 | ENST00000421284.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012596.hg.1 | ENST00000557131.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012596.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR14012596.hg.1 | TC14001117.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1400167281.hg.1 | TC1400009124.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000261699.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000267436.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000421284.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000554191.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555423.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000555610.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | ENST00000556393.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1400009124.hg.1 | NM_024884.2 | Refseq |

L2HGDH
Genomic Map