Assay Details
| Gene Symbol: GLT1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144669.2 | NP_653270.1 | 4 - 5 | 522 | 79 | |
| GenBank mRNA | AK056540.1 | - | 4 - 5 | 420 | 79 | |
More Information
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |

GLT1D1
Genomic Map