Assay Details
| Gene Symbol: GLT1D1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_144669.2 | NP_653270.1 | 5 - 6 | 566 | 68 | |
| NR_133646.1 | 8 - 9 | 754 | 68 | |||
| XM_005253553.4 | XP_005253610.2 | 9 - 10 | 917 | 68 | ||
| XM_011537956.2 | XP_011536258.2 | 10 - 11 | 965 | 68 | ||
| XM_011537957.2 | XP_011536259.1 | 9 - 10 | 654 | 68 | ||
| XM_017018855.1 | XP_016874344.1 | 10 - 11 | 965 | 68 | ||
| XM_017018856.1 | XP_016874345.1 | 8 - 9 | 557 | 68 | ||
| XM_017018857.1 | XP_016874346.1 | 7 - 8 | 530 | 68 | ||
| XM_017018858.1 | XP_016874347.1 | 6 - 7 | 473 | 68 | ||
| XM_017018860.1 | XP_016874349.1 | 6 - 7 | 434 | 68 | ||
| XM_017018861.1 | XP_016874350.1 | 8 - 9 | 599 | 68 | ||
| GenBank mRNA | AK056540.1 | - | 5 - 6 | 464 | 68 | |
| AK295333.1 | - | 10 - 11 | 682 | 68 | ||
| AK295454.1 | - | 7 - 8 | 417 | 68 | ||
| BC043528.1 | - | 8 - 9 | 754 | 5168267 | 68 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC12001027.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR12013542.hg.1 | ENST00000281703.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013542.hg.1 | ENST00000442111.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013542.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013542.hg.1 | ENST00000542193.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013542.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR12013542.hg.1 | TC12001027.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1200169816.hg.1 | TC1200009375.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000281703.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000413816.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000441390.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000442111.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000537468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | ENST00000539044.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200009375.hg.1 | NM_144669.2 | Refseq |

GLT1D1
Genomic Map