Assay Details
| Gene Symbol: COX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001297732.1 | NP_001284661.1 | 1 - 2 | 444 | 78 | |
| NM_173827.3 | NP_776188.1 | 1 - 2 | 444 | 78 | ||
| XM_005265680.4 | XP_005265737.1 | 1 - 2 | 444 | 78 | ||
| XM_017008045.1 | XP_016863534.1 | 1 - 2 | 444 | 78 | ||
| GenBank mRNA | AK096310.2 | - | 1 - 2 | 390 | 78 | |
| AM055750.1 | - | 1 - 2 | 94 | 78 | ||
| AY957564.1 | - | 1 - 2 | 440 | 78 | ||
| AY957565.1 | - | 1 - 2 | 440 | 78 | ||
| BC101684.1 | - | 1 - 2 | 377 | 8069190 | 78 | |
| BC143642.1 | - | 1 - 2 | 377 | 9052156 | 78 | |
| DA385228.1 | - | 1 - 2 | 444 | 78 | ||
| DA674605.1 | - | 1 - 2 | 444 | 78 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04001278.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04017437.hg.1 | TC04001278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017437.hg.1 | ENST00000295890.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017437.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017437.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017441.hg.1 | TC04001278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017441.hg.1 | ENST00000295890.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017441.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017441.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017440.hg.1 | TC04001278.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017440.hg.1 | ENST00000295890.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017440.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017440.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04017440.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000421792.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000449739.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_001300729.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400012930.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | ENST00000507544.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | ENST00000510031.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | NM_001297732.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | NM_001297733.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | NM_173827.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400190227.hg.1 | TC0400012930.hg.1 | Clariom D TC |

COX18
Genomic Map