Assay Details
Gene Symbol: C6orf141 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_005248853.2 | XP_005248910.1 | 1 - 2 | 1029 | 118 | |
GenBank mRNA | BC036917.1 | - | 1 - 2 | 1007 | 5201433 | 118 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000424426.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000424426.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157459.hg.1 | TC0600008212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600157462.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157462.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157462.hg.1 | TC0600008212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000424426.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000424426.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | ENST00000530382.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | TC0600008212.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000415078.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | ENST00000529246.3 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | NM_001145652.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0600157449.hg.1 | TC0600008212.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | ENST00000371194.7 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | ENST00000414696.5 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | ENST00000526429.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0600157460.hg.1 | TC0600008212.hg.1 | Clariom D TC |
