Assay Details
| Gene Symbol: C19orf43 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_024038.2 | NP_076943.1 | 1 - 2 | 407 | 67 | |
| GenBank mRNA | AK027588.1 | - | 1 - 2 | 302 | 67 | |
| BC000216.2 | - | 1 - 2 | 407 | 2961319 | 67 | |
| BC004533.2 | - | 1 - 2 | 352 | 3139656 | 67 | |
| BC015347.1 | - | 1 - 2 | 366 | 4156774 | 67 | |
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Transcript Location | Amplicon Length | |
|---|---|---|---|---|---|---|---|
| GenBankmRNA | BC065537.1 | - | 382 | Chr. - - [-] on Build | 67 | ||
More Information
Set Membership: |
|

C19orf43
Genomic Map