Assay Details
Gene Symbol: OR14C36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001001918.1 | NP_001001918.1 | 1 | 846 | 107 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC01002034.hg.1
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Agilent
Affymetrix
ArrayStar
ABI
Illumina
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006449 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931924 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006464 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006425 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931876 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006465 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931928 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006475 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006477 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006435 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P15205673 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006436 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006466 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006416 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006467 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931897 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006463 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006422 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931922 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006478 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006473 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931870 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006418 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006433 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931912 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006471 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931911 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006479 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931907 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006434 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006441 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006446 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006453 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006423 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006430 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931908 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931883 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006428 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931906 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006427 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006419 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006438 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006437 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006468 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P00931909 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006417 | - | - |
Agilent Human Whole Genome ChIP Tiling Array | A_17_P21006461 | - | - |
Agilent SurePrint G3 Human CNV Microarray 2x400K | 183669 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345464 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0179 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0181 | - | - |
Agilent Genotypic Designed Human Whole genome gene expression profiling microarray 4x44k | GT_HS_44k_0180 | - | - |
Agilent G4851A SurePrint G3 Hmn GE 8x60K Microarray | 14051 | - | - |
Agilent Human Genome CGH + SNP Microarray | A_18_P10592856 | - | - |
Agilent Whole Human Genome Microarray 4x44K v2 | A_33_P3345464 | - | - |
SurePrint G3 Human GE Microarray G4851A | A_33_P3345464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006427 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006436 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006425 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931899 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006453 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006449 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931922 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006445 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006421 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931906 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931870 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006465 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006441 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931908 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931907 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006468 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006416 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931907 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931899 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006423 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931883 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931870 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006446 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P15205673 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006445 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931873 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006434 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006471 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006421 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006464 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006461 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006468 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006436 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006453 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931912 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006465 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006416 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006425 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931911 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006449 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006441 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006433 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006431 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006418 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006435 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006438 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006417 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931897 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006448 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931906 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006422 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006430 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931922 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P00931908 | - | - |
Agilent SurePrint G3 Human Promoter Microarray 1x1M | A_17_P21006427 | - | - |
Agilent Human Genome CGH Microarray 105A (G4412A) | A_16_P00296894 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931898 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931897 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931895 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931885 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931923 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931874 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931909 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931888 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931917 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931890 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931900 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931899 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931883 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931915 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931914 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931903 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931893 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931869 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931880 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931872 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931867 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931906 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931878 | - | - |
Agilent Human Promoter ChIP-on-Chip A) | A_17_P00931921 | - | - |
Agilent Human Genome 244K CGH Microarray (Alpha Test) | A_16_P00296894 | - | - |
Agilent SurePrint G3 CGH array (4X180K) CCA_VUMC design AMADID#022522 | 175409 | - | - |
Agilent HD CGH Microarray (2X105K) Oxford design AMADID#019015 | 104519 | - | - |
Agilent Human CGH Whole Genome Microarray 1x1M (G4447A) | A_16_P00296894 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P15205673 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931923 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931957 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931962 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931914 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931977 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931874 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931890 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931965 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931912 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931869 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931872 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931921 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931967 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931899 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931883 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931867 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931880 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931885 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931930 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931898 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931927 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931953 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931909 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931971 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931954 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931907 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931903 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931959 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931975 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931917 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931924 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931979 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931906 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931895 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931878 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931915 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931893 | - | - |
Agilent Human Whole Genome ChIP-on-Chip | A_17_P00931888 | - | - |
Agilent SurePrint G3 Human CGH Microarray 2x400K | A_16_P00296894 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Gene 1.0 ST Array | NM_001001918_at | - | - |
Affymetrix Human Gene 1.0 ST Array | 7911272 | - | - |
Affymetrix Human Gene 1.1 ST Array | 7911271 | - | - |
Affymetrix Human Genome U219 Array | 11742593_PM_at | - | - |
Affymetrix Human Genome U219 Array | 11742593_at | - | - |
Affymetrix Human Transcriptome Array 2.0 | TC0100012353.hg.1 | ENST00000317861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0100012353.hg.1 | NM_001001918.1 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | PSR01031128.hg.1 | TC01002034.hg.1 | HTA TC |
Affymetrix Human Transcriptome Array 2.0 | PSR01031128.hg.1 | ENST00000317861.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | PSR01031128.hg.1 | NM_001001918.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190631.hg.1 | ENST00000317861.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190631.hg.1 | NM_001001918.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190631.hg.1 | ENST00000317861.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0100190631.hg.1 | NM_001001918.1 | Refseq |
Affymetrix Human Clariom D Assay | PSR0100190631.hg.1 | TC0100012353.hg.1 | Clariom D TC |
Affymetrix Human Clariom D Assay | TC0100012353.hg.1 | ENST00000317861.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0100012353.hg.1 | NM_001001918.1 | Refseq |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042324 | - | - |
Arraystar Human LncRNA microarray V2.0 | ASHG19A3A042324 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
ABI Human Genome Survey Microarray Version 2.0 | 151050 | - | - |
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
HumanHT-12 v4 Expression BeadChip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanWG-6 v2.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
HumanHT-12 V4.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanHT-12 WG-DASL V4.0 R2 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanRef-8 v2 Expression BeadChip | GI_12674-S | - | - |
Illumina human-6 v2.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina humanRef-8 v2.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina Sentrix Human-6 v2 Expression BeadChip | 7000196 | - | - |
Illumina HumanRef-6 v2.0 expression beadchip | ILMN_12674 | - | - |
Illumina HumanRef-8 v3.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanWG-6 v3.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanHT-12 V3.0 expression beadchip | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanWG-6_V2_0_R2 | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
Illumina HumanRef-8 WG-DASL v3.0 | ILMN_1674997 | NM_001001918.1 | human_transcriptome |
