Assay Details
| Gene Symbol: MTBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022045.4 | NP_071328.2 | 16 - 17 | 1950 | 88 | |
| XM_011516962.2 | XP_011515264.1 | 16 - 17 | 1947 | 88 | ||
| GenBank mRNA | BC013136.1 | - | 16 - 17 | 1922 | 4476886 | 88 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000701.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009542.hg.1 | TC08000701.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009542.hg.1 | ENST00000305949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009542.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186043.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186043.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186043.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186043.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186043.hg.1 | TC0800012349.hg.1 | Clariom D TC |

MTBP
Genomic Map