Assay Details
| Gene Symbol: MTBP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_022045.4 | NP_071328.2 | 17 - 18 | 2054 | 109 | |
| GenBank mRNA | BC013136.1 | - | 17 - 18 | 2026 | 4476886 | 109 |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08000701.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08009543.hg.1 | TC08000701.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009543.hg.1 | ENST00000305949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009543.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009544.hg.1 | TC08000701.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009544.hg.1 | ENST00000305949.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08009544.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186044.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186044.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186044.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800186044.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800186044.hg.1 | TC0800012349.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000305949.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000456899.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000519188.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522308.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000522449.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | ENST00000523373.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800012349.hg.1 | NM_022045.4 | Refseq |

MTBP
Genomic Map