Assay Details
| Gene Symbol: SAP30BP | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001301839.1 | NP_001288768.1 | 6 - 7 | 751 | 85 | |
| NM_001301855.1 | NP_001288784.1 | 8 - 9 | 901 | 85 | ||
| NM_013260.7 | NP_037392.1 | 7 - 8 | 799 | 85 | ||
| XM_011524693.2 | XP_011522995.1 | 7 - 8 | 587 | 85 | ||
| XM_011524694.2 | XP_011522996.1 | 6 - 7 | 533 | 85 | ||
| XM_011524695.2 | XP_011522997.1 | 9 - 10 | 822 | 85 | ||
| XM_017024545.1 | XP_016880034.1 | 6 - 7 | 494 | 85 | ||
| XM_017024546.1 | XP_016880035.1 | 8 - 9 | 680 | 85 | ||
| XM_017024547.1 | XP_016880036.1 | 6 - 7 | 491 | 85 | ||
| XM_017024548.1 | XP_016880037.1 | 9 - 10 | 818 | 85 | ||
| XM_017024549.1 | XP_016880038.1 | 9 - 10 | 818 | 85 | ||
| GenBank mRNA | AF087869.1 | - | 7 - 8 | 542 | 85 | |
| AF119664.1 | - | 7 - 8 | 557 | 85 | ||
| AF450482.1 | - | 7 - 8 | 799 | 85 | ||
| AK002202.1 | - | 7 - 8 | 549 | 85 | ||
| AK023157.1 | - | 7 - 8 | 526 | 85 | ||
| AK095265.1 | - | 6 - 7 | 521 | 85 | ||
| AY082382.1 | - | 7 - 8 | 557 | 85 | ||
| BC007592.1 | - | 7 - 8 | 553 | 3346233 | 85 | |
| BC013409.2 | - | 7 - 8 | 529 | 85 | ||
| BC030233.1 | - | 7 - 8 | 559 | 5229398 | 85 | |
| BC063793.1 | - | 7 - 8 | 551 | 6502502 | 85 | |
| BM807177.1 | - | 8 - 9 | 591 | 85 | ||
| BX647066.1 | - | 4 - 5 | 2148 | 85 | ||
More Information
Updated Mapping Information
Set Membership: |
|

SAP30BP
Genomic Map