Assay Details
| Gene Symbol: SYNM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_015286.5 | NP_056101.5 | 2 - 3 | 1057 | 90 | |
| NM_145728.2 | NP_663780.2 | 2 - 3 | 1057 | 90 | ||
| XM_017022035.1 | XP_016877524.1 | 2 - 3 | 1068 | 90 | ||
| GenBank mRNA | AB002351.2 | - | 2 - 3 | 1095 | 90 | |
| AF359284.1 | - | 2 - 3 | 1048 | 90 | ||
| AJ310521.1 | - | 2 - 3 | 1051 | 90 | ||
| AJ310522.1 | - | 2 - 3 | 1051 | 90 | ||
| AJ697971.1 | - | 2 - 3 | 1057 | 90 | ||
| AL833262.1 | - | 2 - 3 | 548 | 90 | ||
| BC151243.1 | - | 2 - 3 | 1102 | 8860516 | 90 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC15000953.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR15008837.hg.1 | ENST00000328642.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008837.hg.1 | ENST00000336292.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008837.hg.1 | ENST00000561323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008837.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008837.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008837.hg.1 | TC15000953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | ENST00000328642.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | ENST00000336292.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | ENST00000560674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | ENST00000561323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008841.hg.1 | TC15000953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008840.hg.1 | ENST00000328642.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008840.hg.1 | ENST00000336292.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008840.hg.1 | ENST00000561323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008840.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008840.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008840.hg.1 | TC15000953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | ENST00000328642.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | ENST00000336292.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | ENST00000560674.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | ENST00000561323.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR15008842.hg.1 | TC15000953.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010604.hg.1 | ENST00000559468.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1500010796.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183722.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183724.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183725.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500183721.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183721.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183721.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183723.hg.1 | TC1500010796.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | ENST00000328642.11 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | ENST00000336292.10 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | ENST00000594047.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | NM_015286.5 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | NM_145728.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1500183726.hg.1 | TC1500010796.hg.1 | Clariom D TC |

SYNM
Genomic Map