Assay Details
| Gene Symbol: MGC32805 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_051996.1 | 4 - 5 | 459 | 134 | ||
| GenBank mRNA | BC029465.1 | - | 4 - 5 | 459 | 134 | |
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0500181766.hg.1 | TC0500011859.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000503529.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000506053.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000509993.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | ENST00000510972.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |
| Affymetrix Human Clariom D Assay | TC0500011859.hg.1 | NR_051996.1 | Refseq |

MGC32805
Genomic Map