Assay Details
| Gene Symbol: CPA6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_020361.4 | NP_065094.3 | 1 - 2 | 375 | 82 | |
| XM_011517570.2 | XP_011515872.1 | 1 - 2 | 914 | 82 | ||
| XM_017013647.1 | XP_016869136.1 | 1 - 2 | 1104 | 82 | ||
| GenBank mRNA | AF221594.1 | - | 1 - 2 | 332 | 82 | |
| AY044833.1 | - | 1 - 2 | 333 | 82 | ||
| BC033684.1 | - | 1 - 2 | 335 | 5193403 | 82 | |
| BI756197.1 | - | 1 - 2 | 375 | 82 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC08001294.hg.1
Additional Information:
This assay is in the 5'UTR region of the AY044833.1,XM_011517570.2 transcripts.
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR08016768.hg.1 | TC08001294.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016768.hg.1 | ENST00000297769.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016768.hg.1 | ENST00000297770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016768.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016768.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016770.hg.1 | TC08001294.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016770.hg.1 | ENST00000297769.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016770.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016767.hg.1 | TC08001294.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016767.hg.1 | ENST00000297769.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016767.hg.1 | ENST00000297770.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016767.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR08016767.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | TC0800010708.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172783.hg.1 | TC0800010708.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172784.hg.1 | TC0800010708.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | ENST00000297770.8 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | ENST00000479862.6 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | ENST00000518549.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172785.hg.1 | TC0800010708.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0800172787.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172787.hg.1 | NM_020361.4 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0800172787.hg.1 | TC0800010708.hg.1 | Clariom D TC |

CPA6
Genomic Map