Assay Details
| Gene Symbol: FCER1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002001.3 | NP_001992.1 | 2 - 3 | 158 | 75 | |
| GenBank mRNA | BC005912.1 | - | 1 - 2 | 81 | 4294467 | 75 |
| BC015195.1 | - | 1 - 2 | 85 | 4251469 | 75 | |
| J03605.1 | - | 1 - 2 | 86 | 75 | ||
| X06948.1 | - | 2 - 3 | 158 | 75 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC01001351.hg.1
Set Membership: |
|

FCER1A
Genomic Map