Assay Details
| Gene Symbol: PRADC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_032319.1 | NP_115695.1 | 2 - 3 | 235 | 63 | |
| GenBank mRNA | AK312077.1 | - | 2 - 3 | 243 | 63 | |
| AY358605.1 | - | 2 - 3 | 226 | 63 | ||
| BC005069.1 | - | 2 - 3 | 235 | 3530601 | 63 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC02001981.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR02031600.hg.1 | TC02001981.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031600.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031600.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031600.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031601.hg.1 | TC02001981.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031601.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031601.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031601.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0300007032.hg.1 | ENST00000412317.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031598.hg.1 | TC02001981.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031598.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031598.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR02031598.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195381.hg.1 | TC0200013061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC0200013061.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195377.hg.1 | TC0200013061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195380.hg.1 | TC0200013061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | ENST00000258083.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | ENST00000470391.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | ENST00000480093.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | NM_032319.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0200195379.hg.1 | TC0200013061.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC0300007032.hg.1 | ENST00000412317.1 | Ensembl |

PRADC1
Genomic Map