Assay Details
| Gene Symbol: PI4KB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_002651.2 | NP_002642.1 | 4 - 5 | 1575 | 88 | |
| XM_011509634.1 | XP_011507936.1 | 4 - 5 | 1575 | 88 | ||
| XM_011509635.1 | XP_011507937.1 | 5 - 6 | 1748 | 88 | ||
| XM_011509636.2 | XP_011507938.1 | 3 - 4 | 1403 | 88 | ||
| XM_011509637.2 | XP_011507939.1 | 4 - 5 | 1576 | 88 | ||
| XM_017001489.1 | XP_016856978.1 | 5 - 6 | 1748 | 88 | ||
| XM_017001490.1 | XP_016856979.1 | 3 - 4 | 1403 | 88 | ||
| XM_017001491.1 | XP_016856980.1 | 4 - 5 | 1576 | 88 | ||
| GenBank mRNA | AB005910.1 | - | 3 - 4 | 1063 | 88 | |
| AJ011121.1 | - | 3 - 4 | 1048 | 88 | ||
More Information
Set Membership: |
|

PI4KB
Genomic Map