Assay Details
| Gene Symbol: NIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016350.4 | NP_057434.4 | 19 - 20 | 2776 | 117 | |
| NM_020921.3 | NP_065972.3 | 20 - 21 | 4915 | 117 | ||
| NM_182944.2 | NP_891989.2 | 20 - 21 | 4915 | 117 | ||
| NM_182946.1 | NP_891991.1 | 20 - 21 | 4915 | 117 | ||
| XM_011536817.1 | XP_011535119.1 | 20 - 21 | 4889 | 117 | ||
| XM_011536819.2 | XP_011535121.1 | 20 - 21 | 4889 | 117 | ||
| XM_011536822.1 | XP_011535124.1 | 19 - 20 | 2750 | 117 | ||
| XM_011536823.2 | XP_011535125.1 | 19 - 20 | 4766 | 117 | ||
| GenBank mRNA | AB046785.2 | - | 19 - 20 | 2698 | 117 | |
| AF186776.1 | - | 1 - 2 | 44 | 117 | ||
| AF212162.2 | - | 19 - 20 | 4944 | 117 | ||
| AF223937.1 | - | 21 - 22 | 4874 | 117 | ||
| AF223938.1 | - | 21 - 22 | 4874 | 117 | ||
| AF223939.1 | - | 21 - 22 | 4874 | 117 | ||
| AF302773.2 | - | 20 - 21 | 4915 | 117 | ||
| AK025652.1 | - | 1 - 2 | 35 | 117 | ||
| AK026083.1 | - | 3 - 4 | 474 | 117 | ||
| AY027794.1 | - | 1 - 2 | 44 | 117 | ||
| AY027795.1 | - | 1 - 2 | 44 | 117 | ||
| AY027796.1 | - | 1 - 2 | 44 | 117 | ||
| AY736182.1 | - | 20 - 21 | 4915 | 117 | ||
| BC090932.1 | - | 17 - 18 | 2620 | 3090109 | 117 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001123.hg.1
Set Membership: |
|

NIN
Genomic Map