Assay Details
| Gene Symbol: NIN | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_016350.4 | NP_057434.4 | 27 - 28 | 3935 | 164 | |
| NM_020921.3 | NP_065972.3 | 28 - 29 | 6074 | 164 | ||
| NM_182944.2 | NP_891989.2 | 28 - 29 | 6074 | 164 | ||
| NM_182946.1 | NP_891991.1 | 28 - 29 | 6074 | 164 | ||
| XM_011536817.1 | XP_011535119.1 | 28 - 29 | 6048 | 164 | ||
| XM_011536819.2 | XP_011535121.1 | 28 - 29 | 6048 | 164 | ||
| XM_011536822.1 | XP_011535124.1 | 27 - 28 | 3909 | 164 | ||
| XM_011536823.2 | XP_011535125.1 | 27 - 28 | 5925 | 164 | ||
| GenBank mRNA | AF186776.1 | - | 9 - 10 | 1203 | 164 | |
| AF212162.2 | - | 27 - 28 | 6103 | 164 | ||
| AF223937.1 | - | 29 - 30 | 6033 | 164 | ||
| AF223938.1 | - | 29 - 30 | 6033 | 164 | ||
| AF223939.1 | - | 29 - 30 | 6033 | 164 | ||
| AF302773.2 | - | 28 - 29 | 6074 | 164 | ||
| AK025652.1 | - | 9 - 10 | 1194 | 164 | ||
| AK026083.1 | - | 11 - 12 | 1633 | 164 | ||
| AY027794.1 | - | 9 - 10 | 1203 | 164 | ||
| AY027795.1 | - | 9 - 10 | 1203 | 164 | ||
| AY027796.1 | - | 9 - 10 | 1203 | 164 | ||
| BC034708.2 | - | 2 - 3 | 256 | 164 | ||
| BC090932.1 | - | 25 - 26 | 3779 | 3090109 | 164 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC14001123.hg.1
Set Membership: |
|

NIN
Genomic Map