Assay Details
Gene Symbol: SLC9A2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
GenBank mRNA | AF073299.1 | - | 6 - 7 | 1605 | 91 | |
BC136377.1 | - | 6 - 7 | 1675 | 9020364 | 91 | |
BC136378.1 | - | 6 - 7 | 1675 | 9020365 | 91 |

More Information
Updated Mapping Information

Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Panther Classification:
MicroArray Probes:
Affymetrix
Array Name | Probe IDs | Targets | Target Type |
---|---|---|---|
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Transcriptome Array 2.0 | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000467657.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | TC0200008680.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | ENST00000233969.2 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | ENST00000469286.1 | Ensembl |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | NM_003048.5 | Refseq |
Affymetrix Human Clariom D Assay | PSR0200160697.hg.1 | TC0200008680.hg.1 | Clariom D TC |
