Assay Details
| Gene Symbol: RABEP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001083585.2 | NP_001077054.1 | 14 - 15 | 2432 | 151 | |
| GenBank mRNA | AF098638.1 | - | 14 - 15 | 2208 | 151 | |
More Information
Set Membership: |
|

RABEP1
Genomic Map