Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008311.hg.1 |
TC10002946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008311.hg.1 |
ENST00000370476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008311.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008312.hg.1 |
TC10002946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008312.hg.1 |
ENST00000370476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008312.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008315.hg.1 |
TC10002946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008315.hg.1 |
ENST00000370476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008315.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008310.hg.1 |
TC10002946.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008310.hg.1 |
ENST00000370476.5 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR10008310.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC1000012492.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186165.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186165.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186165.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186165.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186165.hg.1 |
TC1000012492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186166.hg.1 |
TC1000012492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186167.hg.1 |
TC1000012492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186168.hg.1 |
TC1000012492.hg.1 |
Clariom D TC |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
ENST00000370472.4 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
ENST00000370476.9 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
ENST00000471520.5 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
NM_015960.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR1000186171.hg.1 |
TC1000012492.hg.1 |
Clariom D TC |