Assay Details
| Gene Symbol: GSTT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001293812.1 | NP_001280741.1 | 2 - 3 | 285 | 84 | |
| GenBank mRNA | BX401239.2 | - | 2 - 3 | 233 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001293812.1 transcripts.
Set Membership: |
|

GSTT1
Genomic Map