Assay Details
| Gene Symbol: STX18 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011513483.1 | XP_011511785.1 | 6 - 7 | 613 | 137 | |
| XM_017008296.1 | XP_016863785.1 | 6 - 7 | 613 | 137 | ||
More Information
Additional Information:
Based on assay re-evaluation, this assay may detect transcript(s) from off target gene(s). Please contact Tech Support if you need more information.
Set Membership: |
|

STX18
Genomic Map