Assay Details
Gene Symbol: PC | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_022172.2 | NP_071504.2 | 1 - 2 | 44 | 64 | |
GenBank mRNA | S72370.1 | - | 1 - 2 | 77 | 64 |

More Information
Updated Mapping Information

This assay targets the same location as HTA transcript cluster(s) TC11001976.hg.1
Additional Information:
This assay is in the 5'UTR region of the NM_022172.2,S72370.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
MicroArray Probes:
