Array Name |
Probe IDs |
Targets |
Target Type |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034030.hg.1 |
TC02002105.hg.1 |
HTA TC |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034030.hg.1 |
ENST00000258439.2 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034030.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034030.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
PSR02034030.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000435268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000435268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
ENST00000435268.1 |
Ensembl |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Transcriptome Array 2.0 |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000435268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000435268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
ENST00000435268.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
TC0200013516.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
ENST00000258439.7 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
ENST00000432959.1 |
Ensembl |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
NM_001193304.2 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
NM_017849.3 |
Refseq |
Affymetrix Human Clariom D Assay |
PSR0200198850.hg.1 |
TC0200013516.hg.1 |
Clariom D TC |