Assay Details
| Gene Symbol: DTX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | XM_011538009.2 | XP_011536311.1 | 1 - 2 | 232 | 48 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011538009.2 transcripts.
Set Membership: |
|
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
transcription, DNA-templated
transcription from RNA polymerase II promoter
cell surface receptor signaling pathway
Notch signaling pathway
regulation of Notch signaling pathway
glial cell differentiation
protein ubiquitination
negative regulation of T cell differentiation
negative regulation of neuron differentiation
regulation of nucleic acid-templated transcription
transcription coactivator activity
Notch binding
protein binding
zinc ion binding
ligase activity
SH3 domain binding
ubiquitin protein ligase binding
transcription, DNA-templated
transcription from RNA polymerase II promoter
cell surface receptor signaling pathway
Notch signaling pathway
regulation of Notch signaling pathway
glial cell differentiation
protein ubiquitination
negative regulation of T cell differentiation
negative regulation of neuron differentiation
regulation of nucleic acid-templated transcription
transcription coactivator activity
Notch binding
protein binding
zinc ion binding
ligase activity
SH3 domain binding
ubiquitin protein ligase binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000257600.3 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547730.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000547974.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000548759.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | ENST00000553140.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1200008924.hg.1 | NM_004416.2 | Refseq |

DTX1
Genomic Map