Assay Details
| Gene Symbol: DGKQ | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001347.3 | NP_001338.2 | 7 - 8 | 965 | 64 | |
| XM_011513411.1 | XP_011511713.1 | 7 - 8 | 950 | 64 | ||
| XM_011513412.1 | XP_011511714.1 | 7 - 8 | 950 | 64 | ||
| XM_011513414.2 | XP_011511716.1 | 7 - 8 | 950 | 64 | ||
| XM_011513415.1 | XP_011511717.1 | 7 - 8 | 950 | 64 | ||
| XM_017007814.1 | XP_016863303.1 | 7 - 8 | 950 | 64 | ||
| XM_017007815.1 | XP_016863304.1 | 7 - 8 | 950 | 64 | ||
| GenBank mRNA | AB209503.1 | - | 8 - 9 | 755 | 64 | |
| BC063801.1 | - | 7 - 8 | 973 | 6526640 | 64 | |
| GU727647.1 | - | 7 - 8 | 903 | 64 | ||
| L38707.1 | - | 7 - 8 | 925 | 64 | ||
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC04000958.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Panther Classification:
Gene Ontology Categories:
Function(s)
Process(es)
regulation of transcription from RNA polymerase II promoter
G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
cAMP-mediated signaling
platelet activation
response to ATP
thrombin receptor signaling pathway
protein kinase C signaling
diacylglycerol kinase activity
protein binding
ATP binding
kinase binding
activating transcription factor binding
phospholipase binding
metal ion binding
regulation of transcription from RNA polymerase II promoter
G-protein coupled receptor signaling pathway
protein kinase C-activating G-protein coupled receptor signaling pathway
phosphorylation
cAMP-mediated signaling
platelet activation
response to ATP
thrombin receptor signaling pathway
protein kinase C signaling
diacylglycerol kinase activity
protein binding
ATP binding
kinase binding
activating transcription factor binding
phospholipase binding
metal ion binding
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | PSR04012954.hg.1 | TC04000958.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012954.hg.1 | ENST00000273814.3 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR04012954.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000502309.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000510286.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | ENST00000515182.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | TC0400009712.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | ENST00000273814.7 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | ENST00000509465.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | NM_001347.3 | Refseq |
| Affymetrix Human Clariom D Assay | PSR0400166510.hg.1 | TC0400009712.hg.1 | Clariom D TC |

DGKQ
Genomic Map