Assay Details
| Gene Symbol: LINC00371 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NR_102431.2 | 3 - 4 | 316 | 81 | ||
| NR_102432.2 | 2 - 3 | 385 | 81 | |||
| GenBank mRNA | CX751673.1 | - | 316 | 81 | ||
| DR156254.1 | - | 2 - 3 | 385 | 81 | ||
| DV080333.1 | - | 2 - 3 | 276 | 81 | ||
More Information
Updated Mapping Information
Set Membership: |
|
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1300008973.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000433280.2 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000563710.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | ENST00000569306.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | NR_102431.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | NR_102432.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1300160572.hg.1 | TC1300008973.hg.1 | Clariom D TC |

LINC00371
Genomic Map