Assay Details
| Gene Symbol: CENPV | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_181716.2 | NP_859067.2 | 2 - 3 | 574 | 86 | |
| GenBank mRNA | AF514992.1 | - | 2 - 3 | 510 | 86 | |
| BC052604.1 | - | 2 - 3 | 584 | 5934088 | 86 | |
| BC137486.1 | - | 2 - 3 | 606 | 9021486 | 86 | |
| BC137487.1 | - | 2 - 3 | 606 | 9021487 | 86 | |
More Information
Updated Mapping Information
This assay targets the same location as HTA transcript cluster(s) TC17001173.hg.1
Set Membership: |
|
Matching Ambion siRNAs:
Gene Ontology Categories:
MicroArray Probes:
Affymetrix
| Array Name | Probe IDs | Targets | Target Type |
|---|---|---|---|
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015957.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015957.hg.1 | ENST00000476243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015957.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015957.hg.1 | TC17001173.hg.1 | HTA TC |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015954.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015954.hg.1 | ENST00000476243.1 | Ensembl |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015954.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Transcriptome Array 2.0 | PSR17015954.hg.1 | TC17001173.hg.1 | HTA TC |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700177288.hg.1 | TC1700009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | PSR1700177294.hg.1 | TC1700009888.hg.1 | Clariom D TC |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000299736.4 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000472570.2 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000476243.5 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000482983.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000582062.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000584214.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | ENST00000631687.1 | Ensembl |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |
| Affymetrix Human Clariom D Assay | TC1700009888.hg.1 | NM_181716.2 | Refseq |

CENPV
Genomic Map