Assay Details
| Gene Symbol: TIMP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_003254.2 | NP_003245.1 | 2 - 3 | 311 | 61 | |
| XM_017029766.1 | XP_016885255.1 | 2 - 3 | 265 | 61 | ||
| GenBank mRNA | A10416.1 | - | 2 - 3 | 181 | 61 | |
| AK074854.1 | - | 2 - 3 | 289 | 61 | ||
| AK296116.1 | - | 2 - 3 | 265 | 61 | ||
| AK310206.1 | - | 2 - 3 | 163 | 61 | ||
| AK311937.1 | - | 2 - 3 | 163 | 61 | ||
| BC000866.1 | - | 2 - 3 | 264 | 3460153 | 61 | |
| BC007097.1 | - | 2 - 3 | 212 | 61 | ||
| BU857950.1 | - | 358 | 61 | |||
| CR407638.1 | - | 1 - 2 | 119 | 61 | ||
| CR541982.1 | - | 1 - 2 | 119 | 61 | ||
| M12670.1 | - | 2 - 3 | 167 | 61 | ||
| M59906.1 | - | 2 - 3 | 162 | 61 | ||
| S68252.1 | - | 1 - 2 | 122 | 61 | ||
| X02598.1 | - | 2 - 3 | 160 | 61 | ||
| X03124.1 | - | 2 - 3 | 181 | 61 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the AK296116.1, XM_017029766.1 transcripts.
Set Membership: |
|

TIMP1
Genomic Map