| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
TC05001995.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
ENST00000313708.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
ENST00000318060.7 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
ENST00000380654.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
ENST00000517373.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
ENST00000518836.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR05027727.hg.1 |
NM_024007.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000313708.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000380654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000517373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000518836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000519739.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000519890.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000522192.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000622875.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
NM_001290360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
NM_024007.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
NM_182708.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000313708.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000380654.8 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000517373.1 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000518836.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000519739.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000519890.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000522192.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
ENST00000622875.4 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
NM_001290360.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
NM_024007.4 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
NM_182708.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0500188109.hg.1 |
TC0500012631.hg.1 |
Clariom D TC |