| Array Name |
Probe IDs |
Targets |
Target Type |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
TC0X001396.hg.1 |
HTA TC |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
ENST00000298090.6 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
ENST00000343004.5 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
ENST00000370790.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
ENST00000394270.4 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
ENST00000486347.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
ENST00000493333.1 |
Ensembl |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
NM_001166599.3 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
NM_001166600.2 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
NM_001170756.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
NM_001170757.1 |
Refseq |
| Affymetrix Human Transcriptome Array 2.0 |
PSR0X018590.hg.1 |
NM_145284.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000298090.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000343004.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000370790.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000486347.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000493333.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000611027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001166599.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001166600.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001170756.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001170757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_145284.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000298090.10 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000343004.9 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000370790.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000486347.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000493333.5 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
ENST00000611027.2 |
Ensembl |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001166599.3 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001166600.2 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001170756.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_001170757.1 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
NM_145284.5 |
Refseq |
| Affymetrix Human Clariom D Assay |
PSR0X00173855.hg.1 |
TC0X00010877.hg.1 |
Clariom D TC |